Diethylene glycol monobutyl ether 99+%

Warning

Synonyms: Butyldigol2-(2-Butoxyethoxy)ethanolDiethylene glycol mono-n-butyl ether

AC14971-0025 AC14971-0010
103510-496EA 92.08 USD
103510-496 200001-156
Diethylene glycol monobutyl ether 99+%
Diethylene glycol monobutyl ether
Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 231 °C (760 mmHg)
Melting Pt: –68 °C
Density: 0.955 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 12,01592

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR