Diethylamine 99+%, Extra Pure

Danger

AC14945-0100 AC14945-0010 AC14945-0025 AC14945-2500
103525-720EA 43.1 USD
103525-720 103525-718 200001-108 200006-288
Diethylamine 99+%, Extra Pure
Diethylamine
Formula: (C₂H₅)₂NH
MW: 73.14 g/mol
Boiling Pt: 55 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.71 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 13,03138
https://us.vwr.com/stibo/web/std.lang.all/76/20/2297620.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR