2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone

Danger

Synonyms: Diisobutyl Ketone

AC14944-0025 AC14944-0200 AC14944-0010
200016-220EA 39.1 USD
200016-220 200046-554 200004-918
2,6-Dimethyl-4-heptanone, technical , remainder mainly 4,6-dimethyl-2-heptanone
2,6-Dimethyl-4-heptanone
Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.806 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008940
CAS Number: 108-83-8
EINECS: 203-620-1
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR