4-Ethylmorpholine 99%

Danger

Synonyms: N-Ethylmorpholine

AC14940-5000 AC14940-0025 AC14940-1000
200001-106EA 53.8 USD
200001-106 200007-136 103525-716
4-Ethylmorpholine 99%
4-Ethylmorpholine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 139 °C (760 mmHg)
Melting Pt: –63 °C
Density: 0.9 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00006177
CAS Number: 100-74-3
EINECS: 202-885-0
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR