2-Picoline 98%

Warning

Synonyms: 2-Methylpyridineα-Picoline

AC14933-0010 AC14933-0025 AC14933-0250
200009-578EA 30.7 USD
200009-578 200028-950 200009-576
2-Picoline 98%
2-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 129.5 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.94 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00006332
CAS Number: 109-06-8
EINECS: 203-643-7
UN: 1992
ADR: 3,III
Merck Index: 12,07554

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR