Amyl acetate 99%

Warning

Synonyms: Acetic acid amyl estern-Pentyl acetatePentyl acetateAcetic acid pentyl ester

AC14918-0010 AC14918-2500 AC14918-0025
103525-700EA 198.46 USD
103525-700 200013-564 200032-124
Amyl acetate 99%
Amyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149 °C (760 mmHg)
Melting Pt: –70.8 °C
Density: 0.87 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00009500
CAS Number: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/27/75/722775.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR