1-Bromoadamantane 99%

Synonyms: 1-Adamantyl bromide

AC14903-1000 AC14903-0250
200015-724EA 41.2 USD
200015-724 200008-606
1-Bromoadamantane 99%
1-Bromoadamantane
Formula: C₁₀H₁₅Br
MW: 215.13 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00074721
CAS Number: 768-90-1
EINECS: 212-200-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR