2-Methyl-2-propanethiol 99%

Danger

Synonyms: 2-Methyl-2-propanethioltert-Butyl mercaptantert-Butyl Mercaptantert-Butylthiol

AC14902-0500 AC14902-0025
200005-102EA 20.9 USD
200005-102 200005-100
2-Methyl-2-propanethiol 99%
2-Methyl-2-propanethiol
Formula: C₄H₁₀S
MW: 90.19 g/mol
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –1.1 °C
Density: 0.8 g/cm³
Flash Pt: –24 °C
Storage Temperature: Ambient
MDL Number: MFCD00004857
CAS Number: 75-66-1
EINECS: 200-890-2
UN: 2347
ADR: 3,II
Merck Index: 12,01613

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR