Sodium propionate 99.0-100.5%

Synonyms: SodiumpropionatePropionic acid sodium salt

AC14901-0050 AC14901-0010 AC14901-0250
103516-490EA 222 USD
103516-490 200014-232 200007-496
Sodium propionate 99.0-100.5%
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 285…286 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS Number: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR