Allylthiourea 98%

Danger

Synonyms: 1-Allyl-2-thioureaN-AllylthioureaTiosinamine

AC14880-5000 AC14880-0250 AC14880-1000
200030-324EA 71.8 USD
200030-324 200011-032 200011-034
Allylthiourea 98%
Allylthiourea
Formula: H₂CCHCH₂NHC(S)NH₂
MW: 116.19 g/mol
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: 70…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004940
CAS Number: 109-57-9
EINECS: 203-683-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,09500

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR