4-Bromobenzoic acid 97%

Warning

Synonyms: p-Bromobenzoic Acid

AC14870-1000 AC14870-5000 AC14870-0250
200017-400EA 115 USD
200017-400 103510-426 200005-392
4-Bromobenzoic acid 97%
4-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 251…256 °C
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Merck Index: 13,01392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR