2,4-Xylidine 99%

Danger

Synonyms: 2,4-Dimethylanilinem-Xylidine2,4-Xylidine4-Amino-m-xylene

AC14857-0010 AC14857-1000
200028-282EA 146 USD
200028-282 200008-994
2,4-Xylidine 99%
2,4-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.98 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007738
CAS Number: 95-68-1
EINECS: 202-440-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR