Bis (8-hydroxyquinolinium) sulfate monohydrate 98%

Warning

Synonyms: 8-Hydroxyquinoline hemisulfate salt hemihydrateQuinolin-8-ol hemisulfate hemihydrate8-Hydroxyquinolinium hemisulphate hemihydrate8-Hydroxyquinoline sulphate monohydrate8-Hydroxyquinoline hemisulphate salt hemihydrateQuinolin-8-ol hemisulphate hemihydrate8-Hydroxyquinolinium hemisulfate hemihydrate8-Hydroxyquinoline sulfate monohydrate

AC14845-5000 AC14845-1000
200024-240EA 206 USD
200024-240 200024-238
Bis (8-hydroxyquinolinium) sulfate monohydrate 98%
Bis (8-hydroxyquinolinium) sulphate monohydrate
Formula: C₁₈H₁₆N₂O₆S·1H₂O
MW: 388.4 g/mol
Melting Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00149610
CAS Number: 207386-91-2
EINECS: 205-137-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR