Dibenzyl ether 99%
|
Warning
|
Synonyms:
Benzyl ether
AC14840-0010
AC14840-0025
200011-360EA
54.3
USD
200011-360
200024-524
Dibenzyl ether 99%
Dibenzyl ether
Formula:
C₆H₅CH₂OCH₂C₆H₅ MW: 198.26 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: 1.5…3.5 °C Density: 1.04 g/cm³ Flash Pt: 135 °C |
MDL Number:
MFCD00004780 CAS Number: 103-50-4 EINECS: 203-118-2 UN: 3082 ADR: 9,III Merck Index: 13,01134 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...