α,α,α-Trifluoroacetophenone 99%

Warning

Synonyms: Phenacyl trifluorideTrifluoroacetylbenzene2,2,2-Trifluoroacetophenone1-Phenyl-2,2,2-trifluoro-1-ethanone

AC14835-1000 AC14835-0250 AC14835-0050
103531-294EA 253 USD
103531-294 200018-060 200011-060
α,α,α-Trifluoroacetophenone 99%
α,α,α-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 165…166 °C (760 mmHg)
Melting Pt: –40 °C
Density: 1.24 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00000420
CAS Number: 434-45-7
EINECS: 207-103-1
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/73/27/2297327.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR