α-Bromo-2-nitrotoluene 97%

Danger

Synonyms: 1-(Bromomethyl)-2-nitrobenzenealpha-Bromo-2-nitrotoluene2-Nitrobenzyl bromideα-Bromo-o-nitrotoluene

AC14831-0250 AC14831-0050
200016-726EA 171 USD
200016-726 200022-874
α-Bromo-2-nitrotoluene 97%
α-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 44…48 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR