2-Nitrobenzyl chloride 98%

Danger

Synonyms: 2-Nitrobenzyl chloride1-(Chloromethyl)-2-nitrobenzeneα-Chloro-o-nitrotoluenealpha-Chloro-2-nitrotoluene

AC14816-0100 14816-0100 AC14816-0500
200022-876EA 99.7 USD
200022-876 TS14816-0100 200028-900
2-Nitrobenzyl chloride 98%
2-Nitrobenzyl chloride
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 127…133 °C (10 mmHg)
Melting Pt: 44…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007185
CAS Number: 612-23-7
EINECS: 210-300-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR