3-Bromothiophene 97%

Danger

Synonyms: 3-Thienyl bromide

AC14780-1000 AC14780-0250 AC14780-5000
200009-846EA 187 USD
200009-846 103525-662 200009-848
3-Bromothiophene 97%
3-Bromothiophene
Formula: C₄H₃BrS
MW: 163.04 g/mol
Boiling Pt: 158 °C (760 mmHg)
Density: 1.74 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 3384
ADR: 6.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR