Cyclohexanol 98%

Warning

Synonyms: 1-Hydroxycyclohexane

AC14768-0010 AC14768-0100 AC14768-0025
200001-034EA 51.8 USD
200001-034 200001-036 103510-414
Cyclohexanol 98%
Cyclohexanol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 161 °C (760 mmHg)
Melting Pt: 23 °C
Density: 0.96 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003855
CAS Number: 108-93-0
EINECS: 203-630-6
Merck Index: 13,02754
https://us.vwr.com/stibo/web/std.lang.all/21/52/772152.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR