Triphenyl phosphate ≥99%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: TPP

AC14767-2500 AC14767-0025 AC14767-0050 AC14767-0010
200021-078EA 30.05 USD InStock
200021-078 103525-656 103525-658 200015-044
Triphenyl phosphate ≥99%
Triphenyl phosphate
Formula: C₁₈H₁₅O₄P
MW: 326.29 g/mol
Boiling Pt: 370 °C (760 mmHg)
Melting Pt: 47…53 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00003031
CAS Number: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9,III
Merck Index: 12,09872

Order Now

Specification Test Results

Appearance White Crystalline flakes
Infrared spectrum Conforms
Melting point 47°C to 53°C
Water ≤0.1 %
Additional info Free phenols: 0.005%
GC ≥99.0 %
Color scale ≤50 APHA (after heat treatment)
Acid value ≤0.05 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR