1,1,1,3,3,3-Hexafluoro-2-propanol 99.5+%, pure

Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcoholHFIP1,1,1,3,3,3-Hexafluoropropan-2-olHexafluoro-2-propanolHexafluoroisopropanol

AC14754-0050 AC14754-5000
103525-652EA 972.46 USD
103525-652 200004-590
1,1,1,3,3,3-Hexafluoro-2-propanol 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula: C₃H₂F₆O
MW: 168.04 g/mol
Boiling Pt: 59 °C (760 mmHg)
Melting Pt: –4 °C
Density: 1.596 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,I
Merck Index: 13,09598

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR