Pentafluorobenzoyl chloride ≥98%

Supplier: ACROS ORGANICS MS

Danger

Synonyms: PFBC2,3,4,5,6-Pentafluorobenzoyl chloride

AC14712-1000 AC14712-0050 AC14712-0250
103525-634EA 770.2 USD
103525-634 200026-358 200044-380
Pentafluorobenzoyl chloride ≥98%
Pentafluorobenzoyl chloride
Formula: C₇ClF₅O
MW: 230.52 g/mol
Boiling Pt: 158 °C (1013 hPa)
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000657
CAS Number: 2251-50-5
EINECS: 218-842-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR