Pentafluorobenzenesulfonyl chloride 99%

Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzenesulphonyl chloridePentafluorobenzene-1-sulphonyl chloridePentafluorobenzene-1-sulfonyl chloride

AC14711-0050 AC14711-0010 AC14711-0250
103531-266EA 61.8 USD
103531-266 200013-922 200032-464
Pentafluorobenzenesulfonyl chloride 99%
Pentafluorobenzenesulphonyl chloride
Formula: C₆ClF₅O₂S
MW: 266.58 g/mol
Boiling Pt: 211 °C (760 mmHg)
Density: 1.796 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007427
CAS Number: 832-53-1
EINECS: 212-620-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR