Pentafluorobenzaldehyde 98%

Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzaldehyde

AC14709-0100 AC14709-0500 AC14709-0025
103525-632EA 335 USD
103525-632 200007-280 200020-322
Pentafluorobenzaldehyde 98%
Pentafluorobenzaldehyde
Formula: C₇HF₅O
Boiling Pt: 164…166 °C (760 mmHg)
Melting Pt: 20 °C
Density: 1.588 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00003303
CAS Number: 653-37-2
EINECS: 211-502-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR