4-Nitroanisole 99+%

Warning

AC14702-0050 AC14702-2500
200017-656EA 70.7 USD
200017-656 200017-658
4-Nitroanisole 99+%
4-Nitroanisole
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 51…53 °C
Density: 1.23 g/cm³
Flash Pt: 130 °C
MDL Number: MFCD00007327
CAS Number: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Merck Index: 12,06682

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR