6-Methoxyquinaldine 97%

AC14698-0010
200017-898EA 47.4 USD
200017-898
6-Methoxyquinaldine 97%
6-Methoxyquinaldine
Formula: C₁₁H₁₁NO
MW: 173.21 g/mol
Boiling Pt: 95…96 °C (0.5 mmHg)
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006761
CAS Number: 1078-28-0
EINECS: 214-080-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR