Ethyl-4-fluorobenzoate 99%

Warning

Synonyms: 4-Fluorobenzoic acid ethyl esterEthyl 4-fluorobenzoate

AC14680-1000 AC14680-0250 AC14680-0050
103510-472EA 211 USD
103510-472 200025-308 200025-306
Ethyl-4-fluorobenzoate 99%
Ethyl 4-fluorobenzoate
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 25…27 °C
Density: 1.14 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000351
CAS Number: 451-46-7
EINECS: 207-194-8
Merck Index: 13,04198

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR