6-Fluoro-DL-tryptophan 97%

AC14674-1000
200001-018EA 66 USD
200001-018
6-Fluoro-DL-tryptophan 97%
6-Fluoro-DL-tryptophan
Formula: C₁₁H₁₁FN₂O₂
MW: 222.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005653
CAS Number: 7730-20-3
EINECS: 231-788-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR