1,2,3,4-Tetrahydronaphthalene 98+%, pure

Warning

AC14673-0010 AC14673-0100 AC14673-0025
200056-748EA 245.77 USD
200056-748 103525-624 200056-744
1,2,3,4-Tetrahydronaphthalene 98+%, pure
1,2,3,4-Tetrahydronaphthalene
Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 207 °C (760 mmHg)
Melting Pt: –35 °C
Density: 0.973 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001733
CAS Number: 119-64-2
EINECS: 204-340-2
UN: 3082
ADR: 9,III
Merck Index: 12,09360

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR