3-(Diethylamino)phenol 99%

Warning

Synonyms: 3-DiethylaminophenolN,N-Diethyl-3-hydroxyanilineN,N-Diethyl-3-aminophenol

AC14663-1000
200029-270EA 50.8 USD
200029-270
3-(Diethylamino)phenol 99%
3-(Diethylamino)phenol
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 170 °C (15 mmHg)
Melting Pt: 70…73 °C
Flash Pt: 141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002265
CAS Number: 91-68-9
EINECS: 202-090-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR