Cyclohexanecarboxylic acid 98%

Warning

Synonyms: Hexahydrobenzoic acid

AC14656-5000 AC14656-0010 AC14656-1000 AC14656-0250
200011-830EA 23.1 USD
200011-830 200036-928 200011-828 200048-864
Cyclohexanecarboxylic acid 98%
Cyclohexanecarboxylic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 30…32 °C
Density: 1.03 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001461
CAS Number: 98-89-5
EINECS: 202-711-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR