(Methoxymethyl)triphenylphosphonium chloride 98%

Supplier: ACROS ORGANICS MS

Warning

AC14601-0250 AC14601-5000 AC14601-1000
200008-196EA 56.66 USD
200008-196 103525-612 200008-198
(Methoxymethyl)triphenylphosphonium chloride 98%
(Methoxymethyl)triphenylphosphonium chloride
Formula: C₂₀H₂₀ClOP
MW: 342.8 g/mol
Boiling Pt: 413 °C
Melting Pt: 195…197 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011800
CAS Number: 4009-98-7
EINECS: 223-664-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR