o-Xylene 99%, pure

Danger

Synonyms: 1,2-Dimethylbenzene

AC14099-0025 AC14099-0100 AC14099-0010
103525-608EA 626.54 USD
103525-608 200058-174 200058-172
o-Xylene 99%, pure
o-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 143…145 °C (760 mmHg)
Melting Pt: –25 °C
Density: 0.878 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://us.vwr.com/stibo/web/std.lang.all/85/44/768544.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR