4-Vinylpyridine 95% stabilized

Danger

AC14091-2500 AC14091-0010 AC14091-1000
200001-006EA 92.4 USD
200001-006 103525-604 103525-606
4-Vinylpyridine 95% stabilized
4-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 62…65 °C (15 mmHg)
Melting Pt: –20 °C
Density: 0.989 g/cm³
Flash Pt: 48 °C
Storage Temperature: Freezer
MDL Number: MFCD00006447
CAS Number: 100-43-6
EINECS: 202-852-0
UN: 3073
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR