L(+)-Valine 99%

Synonyms: (S)-(+)-ValineValineH-Val-OH

AC14081-5000 AC14081-1000 AC14081-0250 AC14081-0025
103510-456EA 788 USD
103510-456 103510-458 200000-998 200037-832
L(+)-Valine 99%
L(+)-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 315 °C
Density: 1.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR