Valerophenone 98%

Synonyms: 1-Phenyl-1-pentanone1-Phenylpentan-1-one

AC14080-1000 AC14080-0250 AC14080-0050 AC14080-5000
200033-152EA 16.8 USD
200033-152 200015-132 200021-138 103466-764
Valerophenone 98%
Valerophenone
Formula: C₆H₅C(O)(CH₂)₃CH₃
MW: 162.23 g/mol
Boiling Pt: 105…107 °C (5 mmHg)
Melting Pt: –9 °C
Density: 0.98 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9
EINECS: 213-767-3
https://us.vwr.com/stibo/web/std.lang.all/22/93/762293.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR