1-Undecanol 98%

Warning

Synonyms: 1-Undecyl alcoholDecyl carbinolUndecan-1-olUndecyl alcohol1-Hydroxy undecane

AC14068-5000 AC14068-1000
200027-988EA 63.9 USD
200027-988 200004-808
1-Undecanol 98%
1-Undecanol
Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 146 °C (30 mmHg)
Melting Pt: 11 °C
Density: 0.831 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004751
CAS Number: 112-42-5
EINECS: 203-970-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR