α-Tropolone 98%

Synonyms: Tropolone2-Hydroxytroponeα-Tropolone2-Hydroxycyclohepta-2,4,6-trien-1-one1,2-Tropolone2-Hydroxycyclohepta-2,4,6-trienone

AC14058-0050 AC14058-0250 AC14058-0010
200008-028EA 123 USD
200008-028 200015-094 103516-474
α-Tropolone 98%
α-Tropolone
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 115 °C (12 mmHg)
Melting Pt: 50…54 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004158
CAS Number: 533-75-5
EINECS: 208-577-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR