Trityl chloride 98%

Danger

Synonyms: Trt-ClChlorotriphenylmethaneTriphenylmethyl chlorideTriphenylchloromethane(Chloromethanetriyl)tribenzene

AC14040-5000 AC14040-1000 AC14040-0025
200000-974EA 144 USD
200000-974 103525-594 103529-086
Trityl chloride 98%
Trityl chloride
Formula: C₁₉H₁₅Cl
MW: 278.78 g/mol
Boiling Pt: 230…235 °C (20 mmHg)
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR