2,4,6-Trimethylphenol 99%

Danger

AC14021-0100 AC14021-0500
200028-208EA 32.1 USD
200028-208 103531-048
2,4,6-Trimethylphenol 99%
2,4,6-Trimethylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002235
CAS Number: 527-60-6
EINECS: 208-419-2
UN: 2430
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR