2,3,5-Trimethylphenol 98+%

Danger

AC14019-0050
103531-046EA 24.66 USD
103531-046
2,3,5-Trimethylphenol 98+%
2,3,5-Trimethylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 230…231 °C (760 mmHg)
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002228
CAS Number: 697-82-5
EINECS: 211-806-9
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR