2',4',6'-Trihydroxyacetophenone 98%

Warning

Synonyms: PhloroacetophenoneTHAP2-Acetylphloroglucinol

AC13979-0050 AC13979-0250
200008-940EA 38 USD
200008-940 200004-880
2',4',6'-Trihydroxyacetophenone 98%
2',4',6'-Trihydroxyacetophenone
Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 219…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00149091
CAS Number: 480-66-0
EINECS: 207-556-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR