Benzotrifluoride 99+%

Danger

Synonyms: α,α,α-TrifluorotolueneSolvent BTF

AC13977-0025 AC13977-5000 AC13977-0500
200000-962EA 65.3 USD
200000-962 103525-576 200011-056
Benzotrifluoride 99+%
Benzotrifluoride
Formula: C₇H₅F₃
MW: 146.11 g/mol
Boiling Pt: 102 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.19 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000372
CAS Number: 98-08-8
EINECS: 202-635-0
UN: 2338
ADR: 3,II
Merck Index: 13,01111
https://us.vwr.com/stibo/web/std.lang.all/42/46/764246.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR