n-Tridecane 99+%

Danger

Synonyms: Tridecane

AC13951-0250 AC13951-1000 AC13951-5000
200020-196EA 34 USD
200020-196 103510-430 200000-938
n-Tridecane 99+%
n-Tridecane
Formula: H₃C(CH₂)₁₁CH₃
MW: 184.37 g/mol
Boiling Pt: 234 °C (760 mmHg)
Melting Pt: 5…4 °C
Density: 0.75 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00008979
CAS Number: 629-50-5
EINECS: 211-093-4
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR