2,4,6-Trichloropyrimidine 99%

Warning

AC13949-0250 AC13949-1000
200022-150EA 68.6 USD
200022-150 200004-878
2,4,6-Trichloropyrimidine 99%
2,4,6-Trichloropyrimidine
Formula: C₄HCl₃N₂
MW: 183.42 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 23…25 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006063
CAS Number: 3764-01-0
EINECS: 223-183-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR