2,4,6-Trichlorophenol 98%

Warning

AC13948-1000 AC13948-5000
200004-876EA 38.6 USD
200004-876 103525-560
2,4,6-Trichlorophenol 98%
2,4,6-Trichlorophenol
Formula: C₆H₃Cl₃O
MW: 197.45 g/mol
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 64…68 °C
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002172
CAS Number: 88-06-2
EINECS: 201-795-9
UN: 2020
ADR: 6.1,III
Merck Index: 12,09773

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR