2,4,6-Triaminopyrimidine 97%

Warning

Synonyms: Pyrimidine-2,4,6-triyltriamine

AC13925-0250 AC13925-0050 AC13925-1000
200028-196EA 165 USD
200028-196 103510-402 200022-138
2,4,6-Triaminopyrimidine 97%
2,4,6-Triaminopyrimidine
Formula: C₄H₇N₅
MW: 125.13 g/mol
Melting Pt: 246…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006100
CAS Number: 1004-38-2
EINECS: 213-720-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR