p-Tolylacetic acid 99%

Warning

Synonyms: (4-Methylphenyl)acetic acid4-Tolylacetic acid2-(p-Tolyl)acetic acid

AC13912-0250
200020-538EA 72.6 USD
200020-538
p-Tolylacetic acid 99%
p-Tolylacetic acid
Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 265…267 °C (760 mmHg)
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004353
CAS Number: 622-47-9
EINECS: 210-738-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR