o-Toluic acid 98+%

Warning

Synonyms: 2-Methylbenzoic acid

AC13904-1000 AC13904-0050 AC13904-0010
200013-824EA 24.2 USD
200013-824 200020-254 200007-244
o-Toluic acid 98+%
o-Toluic acid
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 103…105 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002477
CAS Number: 118-90-1
EINECS: 204-284-9
Merck Index: 12,09673

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR