4-Tolualdehyde 97%
|
Warning
|
Synonyms:
4-Methylbenzaldehyd
, p-Tolualdehyde
, 4-Tolylaldehyde
, p-Methylbenzaldehyd
, 4-Methylbenzaldehyde
, p-Tolylaldehyde
AC13900-5000
AC13900-1000
AC13900-0025
200020-526EA
115
USD
200020-526
200007-520
103466-402
4-Tolualdehyde 97%
4-Tolualdehyde
Formula:
C₈H₈O MW: 120.15 g/mol Boiling Pt: 204…205 °C (760 mmHg) Melting Pt: –6 °C Density: 1.015 g/cm³ Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006954 CAS Number: 104-87-0 EINECS: 203-246-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...